Gene Transcripts
- Gene Symbol
- DGKE
- Entrez Gene ID
- 8526
- Gene Name
- diacylglycerol kinase epsilon
- Gene Aliases
- AHUS7, DAGK5, DAGK6, DGK, NPHS7
- Chromosome Location
- Chr. 17: 56834099 - 56869565 on Build GRCh38
- UniGene ID
- Hs.239514
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003647.2 |
NP_003638.1
|
8, 9 |
1386 |
|
XM_011525394.2 |
XP_011523696.1
|
8, 9 |
1364 |
|
XM_011525395.2 |
XP_011523697.1
|
8, 9 |
1371 |
|
XM_011525396.2 |
XP_011523698.1
|
8, 9 |
1373 |
|
XM_017025243.1 |
XP_016880732.1
|
8, 9 |
1753 |
|
GenBank mRNA |
AB209365.1 |
- |
7, 8 |
1036 |
|
AK293039.1 |
- |
8, 9 |
1333 |
|
BC130629.1 |
- |
7, 8 |
1230 |
40146660 |
BC130631.1 |
- |
7, 8 |
1230 |
40146662 |
U49379.1 |
- |
8, 9 |
1293 |
|
View Full Product Details