Gene Transcripts
- Gene Symbol
- RRP1
- Entrez Gene ID
- 8568
- Gene Name
- ribosomal RNA processing 1
- Gene Aliases
- D21S2056E, NNP-1, NOP52, RRP1A
- Chromosome Location
- Chr. 21: 43789537 - 43804102 on Build GRCh38
- UniGene ID
- Hs.110757
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003683.5 |
NP_003674.1
|
5, 6 |
513 |
|
XM_017028485.1 |
XP_016883974.1
|
5, 6 |
504 |
|
GenBank mRNA |
AK223185.1 |
- |
5, 6 |
474 |
|
AK223215.1 |
- |
5, 6 |
470 |
|
AK303000.1 |
- |
5, 6 |
473 |
|
AK313116.1 |
- |
5, 6 |
470 |
|
AL137757.1 |
- |
2, 3 |
534 |
|
BC000380.2 |
- |
5, 6 |
460 |
2820455 |
BC014787.1 |
- |
5, 6 |
435 |
|
BG829556.1 |
- |
|
501 |
|
DA275615.1 |
- |
5, 6 |
515 |
|
U79775.1 |
- |
5, 6 |
458 |
|
View Full Product Details