Gene Transcripts
- Gene Symbol
- DCHS1
- Entrez Gene ID
- 8642
- Gene Name
- dachsous cadherin-related 1
- Gene Aliases
- CDH19, CDH25, CDHR6, FIB1, MVP2, PCDH16, VMLDS1
- Chromosome Location
- Chr. 11: 6621324 - 6655849 on Build GRCh38
- UniGene ID
- Hs.199850
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003737.3 |
NP_003728.1
|
14 |
6064 |
|
GenBank mRNA |
AB053446.1 |
- |
14 |
6067 |
|
View Full Product Details