Gene Transcripts

Gene Symbol
TM4SF5
Entrez Gene ID
9032
Gene Name
transmembrane 4 L six family member 5
Gene Aliases
Chromosome Location
Chr. 17: 4771892 - 4783211 on Build GRCh38
UniGene ID
Hs.184194
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003963.2 NP_003954.2 1 151
GenBank mRNA AF027204.1 - 1 152
BC069519.1 - 1 151 7262226
BC093688.1 - 1 148 7939533
BC117163.1 - 1 148 40125714
CR456925.1 - 1 120
CR541798.1 - 1 120

View Full Product Details