Gene Transcripts
- Gene Symbol
- HGS
- Entrez Gene ID
- 9146
- Gene Name
- hepatocyte growth factor-regulated tyrosine kinase substrate
- Gene Aliases
- HRS
- Chromosome Location
- Chr. 17: 81683932 - 81702121 on Build GRCh38
- UniGene ID
- Hs.661056
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004712.4 |
NP_004703.1
|
20, 21 |
2279 |
|
XM_011525463.2 |
XP_011523765.1
|
20, 21 |
2228 |
|
XM_017025297.1 |
XP_016880786.1
|
21, 22 |
2436 |
|
XM_017025298.1 |
XP_016880787.1
|
19, 20 |
2159 |
|
GenBank mRNA |
AF260566.1 |
- |
19, 20 |
1887 |
|
BC003565.1 |
- |
20, 21 |
2221 |
3544197 |
BT009754.1 |
- |
20, 21 |
2144 |
|
D84064.1 |
- |
20, 21 |
2204 |
|
KU178466.1 |
- |
20, 21 |
2144 |
|
KU178467.1 |
- |
17, 18 |
1910 |
|
U43895.1 |
- |
20, 21 |
2219 |
|
View Full Product Details