Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004255.3 | NP_004246.2 | 2 | 336 | |
GenBank mRNA | AI815421.1 | - | 2 | 350 | |
AK026615.1 | - | 2 | 226 | ||
AK026623.1 | - | 2 | 230 | ||
BC024240.2 | - | 2 | 310 | 4815806 | |
CR407649.1 | - | 2 | 196 | ||
DQ987236.1 | - | 2 | 196 | ||
DQ987237.1 | - | 2 | 196 | ||
M22760.1 | - | 2 | 215 |