Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001256106.2 | NP_001243035.1 | 6 | 1914 | |
NM_001256109.2 | NP_001243038.1 | 6 | 1914 | ||
NM_001256111.2 | NP_001243040.1 | 6 | 1728 | ||
NM_004258.5 | NP_004249.2 | 6 | 1914 | ||
XM_017002846.1 | XP_016858335.1 | 7 | 1867 | ||
XM_017002847.1 | XP_016858336.1 | 7 | 1867 | ||
GenBank mRNA | AK298237.1 | - | 6 | 1703 | |
AK307542.1 | - | 7 | 1914 | ||
BC130327.1 | - | 7 | 1875 | 40146358 | |
BC136484.1 | - | 7 | 1875 | 9020473 | |
BC171768.1 | - | 7 | 1875 | 9054422 | |
Z33642.1 | - | 7 | 1867 |