Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145457.2 | NP_001138929.2 | 3, 4 | 387 | |
NM_001145458.2 | NP_001138930.2 | 3, 4 | 387 | ||
NM_004829.6 | NP_004820.2 | 3, 4 | 387 | ||
XM_011527528.2 | XP_011525830.1 | 3, 4 | 495 | ||
XM_011527529.2 | XP_011525831.1 | 3, 4 | 491 | ||
XM_011527530.2 | XP_011525832.1 | 3, 4 | 483 | ||
GenBank mRNA | AJ001383.1 | - | 3, 4 | 388 | |
AJ006121.1 | - | 3, 4 | 356 | ||
BC064806.1 | - | 3, 4 | 374 | 5218848 |