Gene Transcripts

Gene Symbol
GDF11
Entrez Gene ID
10220
Gene Name
growth differentiation factor 11
Gene Aliases
BMP-11, BMP11
Chromosome Location
Chr. 12: 55743278 - 55757194 on Build GRCh38
UniGene ID
Hs.600883
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005811.3 NP_005802.1 3 997
XM_006719194.3 XP_006719257.1 3 999
GenBank mRNA AF028333.1 - 3 878
AF100907.1 - 3 998

View Full Product Details