Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001006624.1 | NP_001006625.1 | 2 | 268 | |
NM_001006625.1 | NP_001006626.1 | 2 | 268 | ||
NM_006474.4 | NP_006465.3 | 2 | 444 | ||
NM_198389.2 | NP_938203.2 | 2 | 444 | ||
XM_006710295.1 | XP_006710358.1 | 2 | 175 | ||
GenBank mRNA | AB127958.1 | - | 2 | 167 | |
AF030428.1 | - | 2 | 347 | ||
AF390106.1 | - | 2 | 350 | ||
AJ225022.1 | - | 2 | 299 | ||
AK075327.1 | - | 2 | 369 | ||
AK307705.1 | - | 2 | 175 | ||
AK307706.1 | - | 3 | 330 | ||
AK312607.1 | - | 2 | 370 | ||
AY194238.1 | - | 2 | 370 | ||
BC014668.1 | - | 2 | 400 | 4876446 | |
BC022812.1 | - | 2 | 400 | 4875444 | |
BQ212176.1 | - | 2 | 265 | ||
BQ214601.1 | - | 2 | 197 | ||
CA312590.1 | - | 5 | 670 |