Gene Transcripts

Gene Symbol
ATXN2L
Entrez Gene ID
11273
Gene Name
ataxin 2 like
Gene Aliases
A2D, A2LG, A2LP, A2RP
Chromosome Location
Chr. 16: 28823048 - 28837237 on Build GRCh38
UniGene ID
Hs.460499
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308230.1 NP_001295159.1 6 856
NM_007245.3 NP_009176.2 6 856
NM_017492.3 NP_059867.3 6 856
NM_145714.2 NP_663760.1 6 856
NM_148414.2 NP_680780.1 6 856
NM_148415.2 NP_680781.1 6 856
NM_148416.2 NP_680782.1 6 856
XM_005255061.1 XP_005255118.1 6 856
XM_005255062.1 XP_005255119.1 6 856
XM_005255063.3 XP_005255120.1 6 856
XM_005255064.1 XP_005255121.1 6 856
XM_005255065.1 XP_005255122.1 6 856
XM_005255066.1 XP_005255123.1 6 856
XM_005255067.1 XP_005255124.1 6 856
XM_005255068.1 XP_005255125.1 6 856
XM_005255069.1 XP_005255126.1 6 856
XM_005255070.1 XP_005255127.1 6 856
XM_005255071.1 XP_005255128.1 6 856
XM_005255074.1 XP_005255131.1 6 856
XM_005255075.1 XP_005255132.1 6 856
XM_005255076.1 XP_005255133.1 6 856
XM_005255077.1 XP_005255134.1 6 739
XM_006721007.1 XP_006721070.1 6 856
XM_006721008.1 XP_006721071.1 6 856
XM_006721009.1 XP_006721072.1 6 856
XM_006721010.2 XP_006721073.1 6 856
XM_006721011.1 XP_006721074.1 6 856
XM_006721012.3 XP_006721075.1 6 856
XM_011545719.2 XP_011544021.1 6 856
XM_011545721.2 XP_011544023.1 6 856
XM_011545722.1 XP_011544024.1 6 556
XM_017022891.1 XP_016878380.1 6 739
XM_017022892.1 XP_016878381.1 6 556
GenBank mRNA AF034373.1 - 8 958
AJ317970.1 - 6 812
AJ317971.2 - 6 812
AJ317972.2 - 6 812
AJ317973.2 - 6 812
AJ317974.2 - 6 812
AK289981.1 - 6 856
AY188334.1 - 8 958
AY188335.1 - 8 958
AY188336.1 - 8 958
AY188337.1 - 8 958
AY188338.1 - 8 958
BC068012.1 - 7 714
BC082760.1 - 6 652 5812230
BC136584.1 - 6 809 9020574
U70671.1 - 6 486

View Full Product Details