Gene Transcripts

Gene Symbol
MGAT4B
Entrez Gene ID
11282
Gene Name
mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isozyme B
Gene Aliases
GNT-IV, GNT-IVB
Chromosome Location
Chr. 5: 179797597 - 179806952 on Build GRCh38
UniGene ID
Hs.567419
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014275.4 NP_055090.1 7 1101
NM_054013.3 NP_463459.1 6 1541
XM_017008998.1 XP_016864487.1 7 927
GenBank mRNA AB000624.1 - 7 774
AK023137.1 - 6 1541
AK225490.1 - 6 575
AK225492.1 - 6 575
AL162067.1 - 4 208
AY358984.1 - 7 782
BC009464.1 - 5 428
BC051835.1 - 7 977 6196058

View Full Product Details