Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001012478.1 | NP_001012496.1 | 1, 2 | 1100 | |
NM_007279.2 | NP_009210.1 | 1, 2 | 1100 | ||
GenBank mRNA | AB451261.1 | - | 1, 2 | 45 | |
AB451387.1 | - | 1, 2 | 45 | ||
BC008740.2 | - | 1, 2 | 104 | 3346087 | |
BC030574.2 | - | 1, 2 | 1004 | 3687471 | |
X64044.1 | - | 1, 2 | 85 |