Gene Transcripts

Gene Symbol
RAB18
Entrez Gene ID
22931
Gene Name
RAB18, member RAS oncogene family
Gene Aliases
RAB18LI1, WARBM3
Chromosome Location
Chr. 10: 27504174 - 27542237 on Build GRCh38
UniGene ID
Hs.406799
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256410.1 NP_001243339.1 1 212
NM_001256411.1 NP_001243340.1 1 212
NM_001256412.1 NP_001243341.1 1 212
NM_001256415.1 NP_001243344.1 1 212
NM_021252.4 NP_067075.1 1 212
NR_046172.1 1 212
GenBank mRNA AF087860.1 - 1 70
AF136974.1 - 1 126
AF137372.1 - 1 16
AF498950.1 - 1 16
AK001555.1 - 1 16
AK223153.1 - 1 39
AK295443.1 - 1 82
AK297629.1 - 1 82
AK299457.1 - 1 82
AL136734.1 - 1 65
AY574034.1 - 1 101
BC015014.2 - 1 66 3922029
BC029350.1 - 1 82 4819866
BP210053.1 - 1 76
BT009840.1 - 1 16
CN357871.1 - 1 281
CR533455.1 - 1 16

View Full Product Details