Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001323614.1 | NP_001310543.1 | 13 | 2173 | |
NM_001323615.1 | NP_001310544.1 | 12 | 2035 | ||
NM_001323616.1 | NP_001310545.1 | 14 | 2305 | ||
NM_001323617.1 | NP_001310546.1 | 14 | 2140 | ||
NM_015327.2 | NP_056142.2 | 13 | 2112 | ||
XM_017000842.1 | XP_016856331.1 | 9 | 2058 | ||
XM_017000843.1 | XP_016856332.1 | 12 | 2040 | ||
GenBank mRNA | AB029012.2 | - | 13 | 2114 | |
AB085691.1 | - | 13 | 2040 | ||
AL137738.1 | - | 2 | 214 | ||
AY168922.1 | - | 13 | 1966 | ||
AY336728.1 | - | 13 | 1963 | ||
BC007453.2 | - | 2 | 559 | ||
BC038296.1 | - | 13 | 2113 | 4824487 |