Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001323614.1 | NP_001310543.1 | 18 | 2779 | |
NM_001323615.1 | NP_001310544.1 | 18 | 2800 | ||
NM_001323616.1 | NP_001310545.1 | 20 | 3070 | ||
NM_001323617.1 | NP_001310546.1 | 20 | 2905 | ||
NM_015327.2 | NP_056142.2 | 19 | 2877 | ||
XM_017000842.1 | XP_016856331.1 | 15 | 2823 | ||
XM_017000843.1 | XP_016856332.1 | 18 | 2805 | ||
GenBank mRNA | AB029012.2 | - | 19 | 2879 | |
AB085691.1 | - | 19 | 2805 | ||
AK027486.1 | - | 6 | 665 | ||
AL137738.1 | - | 8 | 979 | ||
AY168922.1 | - | 19 | 2731 | ||
AY336728.1 | - | 19 | 2728 | ||
BC007453.2 | - | 8 | 1324 | ||
BC038296.1 | - | 19 | 2878 | 4824487 |