Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001284304.1 | NP_001271233.1 | 2 | 274 | |
NM_001284305.1 | NP_001271234.1 | 2 | 351 | ||
NM_014346.3 | NP_055161.1 | 2 | 274 | ||
NR_104292.1 | 3 | 717 | |||
XM_005261496.1 | XP_005261553.1 | 2 | 270 | ||
XM_011530099.1 | XP_011528401.1 | 2 | 270 | ||
XM_011530100.2 | XP_011528402.1 | 2 | 270 | ||
XM_011530101.2 | XP_011528403.1 | 2 | 270 | ||
XM_011530102.2 | XP_011528404.1 | 2 | 270 | ||
XM_017028742.1 | XP_016884231.1 | 2 | 270 | ||
XM_017028743.1 | XP_016884232.1 | 2 | 270 | ||
XM_017028744.1 | XP_016884233.1 | 3 | 262 | ||
XM_017028745.1 | XP_016884234.1 | 2 | 270 | ||
XM_017028746.1 | XP_016884235.1 | 2 | 270 | ||
XM_017028747.1 | XP_016884236.1 | 2 | 270 | ||
GenBank mRNA | AB449908.1 | - | 2 | 104 | |
AK055591.1 | - | 2 | 262 | ||
AK125705.1 | - | 3 | 717 | ||
AK127653.1 | - | 2 | 351 | ||
AK301445.1 | - | 2 | 252 | ||
AK315779.1 | - | 2 | 236 | ||
BC001292.1 | - | 2 | 207 | 3457393 | |
BC020976.1 | - | 2 | 209 | 3884347 | |
BC029897.1 | - | 2 | 239 | 5173045 | |
CR456412.1 | - | 2 | 125 | ||
DA990679.1 | - | 2 | 270 | ||
HY033225.1 | - | 2 | 275 |