Gene Transcripts

Gene Symbol
PNISR
Entrez Gene ID
25957
Gene Name
PNN interacting serine and arginine rich protein
Gene Aliases
C6orf111, HSPC306, SFRS18, SRrp130, bA98I9.2
Chromosome Location
Chr. 6: 99398051 - 99425345 on Build GRCh38
UniGene ID
Hs.520287
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001322405.1 NP_001309334.1 12 2446
NM_001322406.1 NP_001309335.1 13 2562
NM_001322408.1 NP_001309337.1 11 2366
NM_001322410.1 NP_001309339.1 12 2450
NM_001322412.1 NP_001309341.1 12 2446
NM_001322413.1 NP_001309342.1 11 2370
NM_001322414.1 NP_001309343.1 11 2396
NM_001322415.1 NP_001309344.1 10 2316
NM_015491.2 NP_056306.1 11 2370
NM_032870.3 NP_116259.2 12 2450
NR_136326.1 10 3090
XM_005266912.3 XP_005266969.2 11 2329
XM_017010710.1 XP_016866199.1 11 2393
XM_017010711.1 XP_016866200.1 10 2279
XM_017010712.1 XP_016866201.1 12 3801
XM_017010713.1 XP_016866202.1 12 3797
GenBank mRNA AF161379.1 - 1 230
AF314184.1 - 10 2240
AF314185.1 - 1 642
AL080186.1 - 1 691
BQ000404.1 - 2 566

View Full Product Details