Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286577.1 | NP_001273506.1 | 21 | 4022 | |
NM_015531.5 | NP_056346.3 | 21 | 4022 | ||
XM_011544904.1 | XP_011543206.1 | 21 | 3921 | ||
XM_011544905.1 | XP_011543207.1 | 21 | 3921 | ||
XM_017017510.1 | XP_016872999.1 | 21 | 3921 | ||
XM_017017511.1 | XP_016873000.1 | 20 | 3847 | ||
XM_017017512.1 | XP_016873001.1 | 20 | 3812 | ||
XM_017017513.1 | XP_016873002.1 | 11 | 2018 | ||
XM_017017514.1 | XP_016873003.1 | 21 | 3921 | ||
XM_017017515.1 | XP_016873004.1 | 7 | 1571 | ||
XM_017017516.1 | XP_016873005.1 | 7 | 1141 | ||
GenBank mRNA | AB231764.1 | - | 21 | 3836 | |
AK091397.1 | - | 4 | 597 | ||
AK125894.1 | - | 8 | 1683 | ||
AL833903.1 | - | 2 | 219 | ||
BC035599.1 | - | 1 | 73 | ||
BC110508.2 | - | 8 | 1651 |