Gene Transcripts

Gene Symbol
OR8B8
Entrez Gene ID
26493
Gene Name
olfactory receptor family 8 subfamily B member 8
Gene Aliases
TPCR85
Chromosome Location
Chr. 11: 124440150 - 124441085 on Build GRCh38
UniGene ID
Hs.351822
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012378.1 NP_036510.1 1 252
GenBank mRNA AF238488.1 - 1 252
BC130393.1 - 1 325 40146424
BC130419.1 - 1 325 40146450

View Full Product Details