Gene Transcripts

Gene Symbol
TSPAN16
Entrez Gene ID
26526
Gene Name
tetraspanin 16
Gene Aliases
TM-8, TM4-B, TM4SF16
Chromosome Location
Chr. 19: 11296139 - 11326996 on Build GRCh38
UniGene ID
Hs.579784
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282509.1 NP_001269438.1 3 476
NM_012466.3 NP_036598.1 3 476
NR_104199.1 3 476
XM_011527901.1 XP_011526203.1 3 471
XM_011527902.1 XP_011526204.1 3 471
XM_011527903.2 XP_011526205.1 3 407
GenBank mRNA BC029908.1 - 3 446
BM560276.1 - 3 463
DB457828.1 - 3 466

View Full Product Details