Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001001485.2 | NP_001001485.1 | 3 | 600 | |
NM_001001486.1 | NP_001001486.1 | 2 | 277 | ||
NM_001001487.1 | NP_001001487.1 | 2 | 277 | ||
NM_001199179.1 | NP_001186108.1 | 3 | 506 | ||
NM_001199180.1 | NP_001186109.1 | 2 | 495 | ||
NM_001199181.1 | NP_001186110.1 | 2 | 495 | ||
NM_001199182.1 | NP_001186111.1 | 2 | 495 | ||
NM_001199183.1 | NP_001186112.1 | 2 | 368 | ||
NM_001199184.1 | NP_001186113.1 | 2 | 368 | ||
NM_001199185.1 | NP_001186114.1 | 2 | 419 | ||
NM_014382.3 | NP_055197.2 | 2 | 419 | ||
XM_005247354.2 | XP_005247411.1 | 2 | 520 | ||
XM_005247355.2 | XP_005247412.1 | 3 | 647 | ||
XM_005247356.2 | XP_005247413.1 | 3 | 400 | ||
XM_005247358.2 | XP_005247415.1 | 2 | 224 | ||
XM_017006164.1 | XP_016861653.1 | 3 | 308 | ||
GenBank mRNA | AB037768.1 | - | 3 | 507 | |
AF181120.1 | - | 2 | 332 | ||
AF181121.1 | - | 2 | 332 | ||
AF189723.2 | - | 2 | 251 | ||
AF225981.1 | - | 2 | 251 | ||
AK296470.1 | - | 2 | 422 | ||
AK299945.1 | - | 2 | 251 | ||
AK304175.1 | - | 2 | 224 | ||
AK304374.1 | - | 2 | 251 | ||
AK314342.1 | - | 2 | 281 | ||
AY268374.1 | - | 2 | 226 | ||
AY268375.1 | - | 2 | 226 | ||
BC028139.1 | - | 2 | 289 | 5215224 | |
DC343471.1 | - | 2 | 419 | ||
DC345627.1 | - | 2 | 368 | ||
DC422788.1 | - | 2 | 495 |