Gene Transcripts
- Gene Symbol
- KLHL20
- Entrez Gene ID
- 27252
- Gene Name
- kelch like family member 20
- Gene Aliases
- KHLHX, KLEIP, KLHLX
- Chromosome Location
- Chr. 1: 173714912 - 173786826 on Build GRCh38
- UniGene ID
- Hs.495035
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014458.3 |
NP_055273.2
|
11, 12 |
1915 |
|
XM_005245093.3 |
XP_005245150.1
|
12, 13 |
2064 |
|
XM_011509420.2 |
XP_011507722.1
|
10, 11 |
1800 |
|
XM_017001052.1 |
XP_016856541.1
|
11, 12 |
2194 |
|
XM_017001053.1 |
XP_016856542.1
|
10, 11 |
2050 |
|
XM_017001054.1 |
XP_016856543.1
|
11, 12 |
1920 |
|
GenBank mRNA |
AB026190.1 |
- |
11, 12 |
1856 |
|
AJ844466.1 |
- |
10, 11 |
1736 |
|
AK001430.1 |
- |
11, 12 |
1872 |
|
AK300755.1 |
- |
10, 11 |
1312 |
|
AK308048.1 |
- |
10, 11 |
1831 |
|
BC063418.1 |
- |
11, 12 |
1845 |
5221643 |
View Full Product Details