Gene Transcripts
- Gene Symbol
- SNX10
- Entrez Gene ID
- 29887
- Gene Name
- sorting nexin 10
- Gene Aliases
- OPTB8
- Chromosome Location
- Chr. 7: 26291895 - 26374330 on Build GRCh38
- UniGene ID
- Hs.741316
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001199835.1 |
NP_001186764.1
|
4, 5 |
485 |
|
NM_001199837.1 |
NP_001186766.1
|
2, 3 |
290 |
|
NM_001199838.1 |
NP_001186767.1
|
1, 2 |
189 |
|
NM_001318198.1 |
NP_001305127.1
|
5, 6 |
559 |
|
NM_001318199.1 |
NP_001305128.1
|
4, 5 |
360 |
|
NM_013322.2 |
NP_037454.2
|
4, 5 |
438 |
|
XM_006715712.2 |
XP_006715775.1
|
4, 5 |
638 |
|
XM_017012085.1 |
XP_016867574.1
|
6, 7 |
653 |
|
XM_017012086.1 |
XP_016867575.1
|
6, 7 |
1092 |
|
XM_017012087.1 |
XP_016867576.1
|
5, 6 |
582 |
|
XM_017012088.1 |
XP_016867577.1
|
5, 6 |
529 |
|
GenBank mRNA |
AF121860.1 |
- |
4, 5 |
351 |
|
AK296142.1 |
- |
5, 6 |
389 |
|
AK307905.1 |
- |
2, 3 |
239 |
|
AK309162.1 |
- |
1, 2 |
189 |
|
AK312850.1 |
- |
4, 5 |
360 |
|
AK316088.1 |
- |
5, 6 |
535 |
|
BC031050.1 |
- |
4, 5 |
332 |
5278015 |
BC034992.1 |
- |
4, 5 |
411 |
4827599 |
BG399132.1 |
- |
1, 2 |
203 |
|
BJ995940.1 |
- |
2, 3 |
290 |
|
BM147978.1 |
- |
4, 5 |
479 |
|
DA377368.1 |
- |
4, 5 |
360 |
|
View Full Product Details