Gene Transcripts

Gene Symbol
PKN3
Entrez Gene ID
29941
Gene Name
protein kinase N3
Gene Aliases
UTDP4-1
Chromosome Location
Chr. 9: 128702497 - 128720920 on Build GRCh38
UniGene ID
Hs.300485
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317926.1 NP_001304855.1 17 2434
NM_013355.4 NP_037487.2 17 2434
XM_005251946.3 XP_005252003.1 17 2989
XM_006717080.2 XP_006717143.1 17 2503
XM_017014649.1 XP_016870138.1 17 2505
XM_017014650.1 XP_016870139.1 17 2643
GenBank mRNA AB019692.1 - 17 2022
BC062558.1 - 17 2037 6649806

View Full Product Details