Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001077180.2 | NP_001070648.1 | 4 | 866 | |
NM_001288659.1 | NP_001275588.1 | 4 | 523 | ||
NM_001288660.1 | NP_001275589.1 | 4 | 523 | ||
NM_016025.4 | NP_057109.3 | 4 | 866 | ||
XM_017023269.1 | XP_016878758.1 | 3 | 559 | ||
GenBank mRNA | AF151839.1 | - | 4 | 530 | |
AJ278577.1 | - | 4 | 784 | ||
AJ278578.1 | - | 4 | 784 | ||
AK025565.1 | - | 2 | 97 | ||
AK074529.1 | - | 4 | 765 | ||
AK075022.1 | - | 4 | 752 | ||
AK075237.1 | - | 4 | 735 | ||
AK313077.1 | - | 4 | 750 | ||
BC000195.1 | - | 4 | 751 | 3350598 | |
BG183771.1 | - | 4 | 522 |