Gene Transcripts
- Gene Symbol
- CRIM1
- Entrez Gene ID
- 51232
- Gene Name
- cysteine rich transmembrane BMP regulator 1
- Gene Aliases
- CRIM-1, S52
- Chromosome Location
- Chr. 2: 36356088 - 36551135 on Build GRCh38
- UniGene ID
- Hs.699247
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016441.2 |
NP_057525.1
|
8, 9 |
1578 |
|
XM_005264357.4 |
XP_005264414.1
|
7, 8 |
1400 |
|
XM_011532898.2 |
XP_011531200.1
|
9, 10 |
1699 |
|
XM_011532899.2 |
XP_011531201.1
|
9, 10 |
1700 |
|
XM_011532901.2 |
XP_011531203.1
|
9, 10 |
1700 |
|
XM_017004258.1 |
XP_016859747.1
|
8, 9 |
1456 |
|
XM_017004259.1 |
XP_016859748.1
|
8, 9 |
1717 |
|
XM_017004260.1 |
XP_016859749.1
|
7, 8 |
1331 |
|
XM_017004261.1 |
XP_016859750.1
|
7, 8 |
991 |
|
XM_017004262.1 |
XP_016859751.1
|
6, 7 |
874 |
|
GenBank mRNA |
AB209139.1 |
- |
8, 9 |
1578 |
|
AF167706.1 |
- |
8, 9 |
1551 |
|
AY358371.1 |
- |
8, 9 |
1879 |
|
BC111989.1 |
- |
8, 9 |
1549 |
8327457 |
BC113371.1 |
- |
8, 9 |
1549 |
8322423 |
BX648755.1 |
- |
2, 3 |
227 |
|
View Full Product Details