Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016628.4 | NP_057712.2 | 9 | 1343 | |
NM_100264.2 | NP_567822.1 | 9 | 1634 | ||
NM_100486.3 | NP_567823.1 | 8 | 1034 | ||
XM_011519491.2 | XP_011517793.1 | 9 | 1944 | ||
XM_017016315.1 | XP_016871804.1 | 9 | 1454 | ||
XM_017016316.1 | XP_016871805.1 | 9 | 1944 | ||
XM_017016317.1 | XP_016871806.1 | 8 | 1145 | ||
XM_017016318.1 | XP_016871807.1 | 8 | 1145 | ||
GenBank mRNA | AB058747.1 | - | 7 | 1044 | |
AF208858.1 | - | 2 | 269 | ||
AK055852.1 | - | 9 | 1451 | ||
AK223600.1 | - | 9 | 1341 | ||
AK225846.1 | - | 9 | 1447 | ||
AK290174.1 | - | 9 | 1348 | ||
AK297460.1 | - | 10 | 1206 | ||
AL713727.1 | - | 7 | 1473 | ||
BC004258.1 | - | 9 | 1334 | 3347345 | |
BC010356.1 | - | 4 | 485 |