Gene Transcripts

Gene Symbol
AMOTL2
Entrez Gene ID
51421
Gene Name
angiomotin like 2
Gene Aliases
LCCP
Chromosome Location
Chr. 3: 134355345 - 134375417 on Build GRCh38
UniGene ID
Hs.426312
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278683.1 NP_001265612.1 7 2007
NM_001278685.1 NP_001265614.1 7 2015
NM_016201.3 NP_057285.3 7 2021
XM_006713654.2 XP_006713717.1 7 2039
XM_011512881.1 XP_011511183.1 7 1878
XM_017006580.1 XP_016862069.1 7 1878
XM_017006581.1 XP_016862070.1 7 2059
XM_017006582.1 XP_016862071.1 7 1886
GenBank mRNA AB023206.1 - 7 1954
AF175966.1 - 6 1088
AK074731.1 - 2 419
AK091040.1 - 4 1089
AK223598.1 - 7 1873
AK291616.1 - 7 1965
AK299270.1 - 7 2007
BC011454.1 - 5 987 4181482
BC025981.2 - 4 577

View Full Product Details