Gene Transcripts

Gene Symbol
HPCAL4
Entrez Gene ID
51440
Gene Name
hippocalcin like 4
Gene Aliases
HLP4
Chromosome Location
Chr. 1: 39678648 - 39691710 on Build GRCh38
UniGene ID
Hs.743975
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282396.1 NP_001269325.1 3 423
NM_001282397.1 NP_001269326.1 2 317
NM_016257.3 NP_057341.1 2 542
XM_017001447.1 XP_016856936.1 2 155
GenBank mRNA AB001105.1 - 2 524
AK294813.1 - 2 299
AK312314.1 - 2 264
AL136591.1 - 2 249
BC030827.1 - 2 246 4811834
DA220121.1 - 3 349
DC333158.1 - 2 274

View Full Product Details