Gene Transcripts

Gene Symbol
DTL
Entrez Gene ID
51514
Gene Name
denticleless E3 ubiquitin protein ligase homolog
Gene Aliases
CDT2, DCAF2, L2DTL, RAMP
Chromosome Location
Chr. 1: 212035553 - 212105006 on Build GRCh38
UniGene ID
Hs.656473
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286229.1 NP_001273158.1 9 1051
NM_001286230.1 NP_001273159.1 10 1155
NM_016448.3 NP_057532.3 11 1281
XM_011509614.1 XP_011507916.1 11 985
GenBank mRNA AF195765.1 - 11 1045
AF345896.1 - 11 1066
AK000742.1 - 11 1099
AK001261.1 - 2 88
AK027651.1 - 9 758
AK292343.1 - 11 1153
AK303346.1 - 10 958
BC033297.1 - 11 1022 4342989
BC033540.1 - 11 1063 4826434
DQ641253.1 - 11 943

View Full Product Details