Gene Transcripts
- Gene Symbol
- CPSF3
- Entrez Gene ID
- 51692
- Gene Name
- cleavage and polyadenylation specific factor 3
- Gene Aliases
- CPSF-73, CPSF73
- Chromosome Location
- Chr. 2: 9423537 - 9473110 on Build GRCh38
- UniGene ID
- Hs.515972
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001321833.1 |
NP_001308762.1
|
16, 17 |
2314 |
|
NM_001321834.1 |
NP_001308763.1
|
16, 17 |
3222 |
|
NM_001321835.1 |
NP_001308764.1
|
16, 17 |
1957 |
|
NM_001321836.1 |
NP_001308765.1
|
17, 18 |
2404 |
|
NM_016207.3 |
NP_057291.1
|
16, 17 |
2085 |
|
GenBank mRNA |
AF171877.1 |
- |
16, 17 |
1863 |
|
AK025440.1 |
- |
14, 15 |
1744 |
|
AK223487.1 |
- |
16, 17 |
1883 |
|
AK298918.1 |
- |
14, 15 |
1678 |
|
BC011654.2 |
- |
16, 17 |
1958 |
4304135 |
BC014106.1 |
- |
16, 17 |
2875 |
|
BC020211.1 |
- |
16, 17 |
1883 |
4890593 |
EF036508.1 |
- |
3, 4 |
245 |
|
View Full Product Details