Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016848.5 | NP_058544.3 | 4 | 949 | |
XM_011518785.2 | XP_011517087.1 | 4 | 4479 | ||
XM_011518786.2 | XP_011517088.1 | 5 | 328 | ||
XM_017014810.1 | XP_016870299.1 | 4 | 4490 | ||
GenBank mRNA | BC026314.1 | - | 4 | 950 | |
D84361.1 | - | 4 | 822 | ||
KU176125.1 | - | 4 | 246 |