Gene Transcripts

Gene Symbol
DRAM1
Entrez Gene ID
55332
Gene Name
DNA damage regulated autophagy modulator 1
Gene Aliases
DRAM
Chromosome Location
Chr. 12: 101877327 - 101923623 on Build GRCh38
UniGene ID
Hs.525634
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018370.2 NP_060840.2 4 887
XM_005269004.2 XP_005269061.1 4 772
XM_017019578.1 XP_016875067.1 5 1009
GenBank mRNA AK002121.1 - 2 193
AK094923.1 - 4 517
BC013773.1 - 2 185
BC018435.1 - 4 863 4474297

View Full Product Details