Gene Transcripts

Gene Symbol
CDCA7L
Entrez Gene ID
55536
Gene Name
cell division cycle associated 7 like
Gene Aliases
JPO2, R1, RAM2
Chromosome Location
Chr. 7: 21900899 - 21945924 on Build GRCh38
UniGene ID
Hs.744069
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127370.2 NP_001120842.1 3 211
NM_001127371.2 NP_001120843.1 2 157
NM_018719.4 NP_061189.2 2 157
GenBank mRNA AK022955.1 - 2 132
AK095965.1 - 3 165
AK292490.1 - 2 132
AK300781.1 - 2 117
AY161168.1 - 2 37
AY161169.1 - 2 37
BC009352.2 - 2 49 4139111
BC025242.1 - 2 114 4842169
BC032576.1 - 2 125 5491780
BC039823.1 - 2 121 6057601
BX422040.2 - 2 105
DC331792.1 - 2 157

View Full Product Details