Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001174081.1 | NP_001167552.1 | 6 | 1723 | |
NM_001174082.1 | NP_001167553.1 | 5 | 1688 | ||
NM_001174083.1 | NP_001167554.1 | 7 | 1685 | ||
NM_017852.3 | NP_060322.1 | 6 | 1754 | ||
GenBank mRNA | AB209300.1 | - | 5 | 1606 | |
AF298547.1 | - | 5 | 1605 | ||
AF310106.1 | - | 6 | 1699 | ||
AF464764.1 | - | 6 | 1654 | ||
AK000517.1 | - | 6 | 1699 | ||
AK025952.1 | - | 6 | 1663 | ||
AK223253.1 | - | 6 | 1663 | ||
AK223269.1 | - | 6 | 1663 | ||
AK292681.1 | - | 6 | 1682 | ||
AK302989.1 | - | 7 | 1654 | ||
BC001039.2 | - | 5 | 1634 | 3504594 | |
BC003592.1 | - | 6 | 1678 | 2821428 | |
BC039269.1 | - | 6 | 1723 | 5298283 |