Gene Transcripts

Gene Symbol
BRK1
Entrez Gene ID
55845
Gene Name
BRICK1, SCAR/WAVE actin nucleating complex subunit
Gene Aliases
C3orf10, HSPC300, MDS027, hHBrk1
Chromosome Location
Chr. 3: 10115649 - 10127190 on Build GRCh38
UniGene ID
Hs.649307
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018462.4 NP_060932.2 2 230
GenBank mRNA AF161418.1 - 2 191
AF220047.1 - 2 959
AK312155.1 - 2 204
AY148219.1 - 2 230
AY148220.1 - 2 230
BC001067.1 - 2 206 2823162
BC007929.1 - 2 199 3688264
BC019303.1 - 2 197 2823162
BC061515.1 - 2 204

View Full Product Details