Gene Transcripts

Gene Symbol
TEX14
Entrez Gene ID
56155
Gene Name
testis expressed 14, intercellular bridge forming factor
Gene Aliases
CT113
Chromosome Location
Chr. 17: 58556677 - 58692055 on Build GRCh38
UniGene ID
Hs.390221
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001201457.1 NP_001188386.1 6 704
NM_031272.4 NP_112562.3 6 704
NM_198393.3 NP_938207.2 6 704
XM_011525029.2 XP_011523331.1 6 683
XM_017024860.1 XP_016880349.1 5 468
XM_017024861.1 XP_016880350.1 6 708
GenBank mRNA AK125896.1 - 6 694
AK128217.1 - 2 139
AK308299.1 - 6 664
AL834143.1 - 6 703
BC040526.1 - 6 672 4831308

View Full Product Details