Gene Transcripts

Gene Symbol
INTS12
Entrez Gene ID
57117
Gene Name
integrator complex subunit 12
Gene Aliases
INT12, PHF22, SBBI22
Chromosome Location
Chr. 4: 105682627 - 105708725 on Build GRCh38
UniGene ID
Hs.480454
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142471.1 NP_001135943.1 7 929
NM_020395.3 NP_065128.2 8 1091
XM_005263148.4 XP_005263205.1 7 1584
XM_011532143.2 XP_011530445.1 8 1740
XM_011532145.2 XP_011530447.1 9 1227
GenBank mRNA AF242524.1 - 7 923
AK314939.1 - 7 929
BC014442.1 - 8 1039 4866321

View Full Product Details