Gene Transcripts

Gene Symbol
SPRYD7
Entrez Gene ID
57213
Gene Name
SPRY domain containing 7
Gene Aliases
C13orf1, CLLD6
Chromosome Location
Chr. 13: 49912702 - 49936501 on Build GRCh38
UniGene ID
Hs.44235
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127482.2 NP_001120954.1 2, 3 550
NM_020456.3 NP_065189.1 3, 4 667
NR_023351.2 1, 2 383
XM_017020677.1 XP_016876166.1 3, 4 667
GenBank mRNA AF052154.1 - 3, 4 466
AF055016.1 - 3, 4 466
AF334405.1 - 3, 4 506
AK290576.1 - 2, 3 389
AK308818.1 - 3, 4 500
AK316576.1 - 3, 4 473
BC022519.1 - 2, 3 356 4792366
DA388141.1 - 1, 2 208

View Full Product Details