Gene Transcripts
- Gene Symbol
- RBM25
- Entrez Gene ID
- 58517
- Gene Name
- RNA binding motif protein 25
- Gene Aliases
- NET52, RED120, RNPC7, S164, Snu71, fSAP94
- Chromosome Location
- Chr. 14: 73058436 - 73121675 on Build GRCh38
- UniGene ID
- Hs.531106
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021239.2 |
NP_067062.1
|
3, 4 |
359 |
|
XM_011537044.2 |
XP_011535346.1
|
4, 5 |
461 |
|
XM_011537045.2 |
XP_011535347.1
|
2, 3 |
166 |
|
GenBank mRNA |
AK026107.1 |
- |
3, 4 |
339 |
|
AK094697.1 |
- |
2, 3 |
156 |
|
AL832314.1 |
- |
3, 4 |
330 |
|
BC062440.1 |
- |
3, 4 |
363 |
|
BC083496.1 |
- |
3, 4 |
325 |
|
BC111066.1 |
- |
3, 4 |
323 |
|
BC113389.1 |
- |
2, 3 |
139 |
|
BC113391.1 |
- |
2, 3 |
139 |
|
BC136775.1 |
- |
3, 4 |
196 |
9020765 |
BC136776.1 |
- |
3, 4 |
196 |
9020766 |
BC144407.1 |
- |
4, 5 |
264 |
9052928 |
BX647116.1 |
- |
3, 4 |
312 |
|
View Full Product Details