Gene Transcripts

Gene Symbol
DNAI2
Entrez Gene ID
64446
Gene Name
dynein axonemal intermediate chain 2
Gene Aliases
CILD9, DIC2
Chromosome Location
Chr. 17: 74274215 - 74314884 on Build GRCh38
UniGene ID
Hs.147472
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172810.1 NP_001166281.1 5 652
NM_023036.4 NP_075462.3 5 655
XM_011525125.2 XP_011523427.1 5 672
GenBank mRNA AF250288.1 - 5 612
AJ295276.1 - 4 851
BC033013.1 - 4 529 4826432
BC039582.1 - 5 642 5752153
BG723530.1 - 4 529
BI462861.1 - 5 650
BI464166.1 - 5 638

View Full Product Details