Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001042573.2 | NP_001036038.1 | 3 | 311 | |
XM_006722019.3 | XP_006722082.1 | 3 | 436 | ||
XM_017024961.1 | XP_016880450.1 | 2 | 153 | ||
GenBank mRNA | AF512564.1 | - | 3 | 303 | |
AK025518.1 | - | 3 | 311 | ||
AK301115.1 | - | 3 | 309 |