Gene Transcripts

Gene Symbol
PHF23
Entrez Gene ID
79142
Gene Name
PHD finger protein 23
Gene Aliases
hJUNE-1b
Chromosome Location
Chr. 17: 7235028 - 7239506 on Build GRCh38
UniGene ID
Hs.644724
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001284517.1 NP_001271446.1 4 680
NM_001284518.1 NP_001271447.1 4 765
NM_024297.2 NP_077273.2 4 881
GenBank mRNA AK026537.1 - 4 825
AK074766.1 - 4 824
AK122791.1 - 3 797
AK122901.1 - 4 765
AK297040.1 - 4 613
BC002509.2 - 4 829 3051214
BC008630.1 - 1 391
EF071959.1 - 4 827

Gene Symbol
NUP98
Entrez Gene ID
4928
Gene Name
nucleoporin 98
Gene Aliases
ADIR2, NUP196, NUP96
Chromosome Location
Chr. 11: 3675010 - 3797792 on Build GRCh38
UniGene ID
Hs.524750
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA KU847959.1 - 2037

View Full Product Details