Gene Transcripts

Gene Symbol
FSD1
Entrez Gene ID
79187
Gene Name
fibronectin type III and SPRY domain containing 1
Gene Aliases
GLFND, MIR1
Chromosome Location
Chr. 19: 4304594 - 4323846 on Build GRCh38
UniGene ID
Hs.28144
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024333.2 NP_077309.1 9 1054
XM_005259644.2 XP_005259701.1 9 1053
GenBank mRNA AF316829.1 - 9 945
AK021750.1 - 9 954
AK315661.1 - 9 1048
AY032617.1 - 9 977
BC003124.1 - 9 954 3502614
BC016442.1 - 3 658

View Full Product Details