Gene Transcripts

Gene Symbol
OR4K5
Entrez Gene ID
79317
Gene Name
olfactory receptor family 4 subfamily K member 5
Gene Aliases
OR14-16
Chromosome Location
Chr. 14: 19920607 - 19921578 on Build GRCh38
UniGene ID
Hs.554585
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005483.1 NP_001005483.1 1 280
GenBank mRNA BC140947.1 - 1 305

View Full Product Details