Gene Transcripts

Gene Symbol
RHBDF2
Entrez Gene ID
79651
Gene Name
rhomboid 5 homolog 2
Gene Aliases
RHBDL5, RHBDL6, TEC, TOC, TOCG, iRhom2
Chromosome Location
Chr. 17: 76470893 - 76501440 on Build GRCh38
UniGene ID
Hs.464157
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005498.3 NP_001005498.2 10 1495
NM_024599.5 NP_078875.4 11 1657
XM_005257669.3 XP_005257726.1 11 2018
XM_005257670.1 XP_005257727.1 11 1527
XM_011525249.2 XP_011523551.1 11 2974
XM_011525250.2 XP_011523552.1 12 1684
XM_011525251.2 XP_011523553.1 11 1501
XM_017025079.1 XP_016880568.1 11 1931
GenBank mRNA AK025994.1 - 6 681
AK074034.1 - 9 1962
AK225684.1 - 6 634
AK292135.1 - 11 1650
AK292166.1 - 10 1450
AK303434.1 - 9 1311
AL833680.1 - 5 550
AY500247.1 - 10 1346
BC016034.1 - 11 1637 3910543
BC035829.1 - 6 743

View Full Product Details