Gene Transcripts

Gene Symbol
ELMO3
Entrez Gene ID
79767
Gene Name
engulfment and cell motility 3
Gene Aliases
CED-12, CED12, ELMO-3
Chromosome Location
Chr. 16: 67199125 - 67204024 on Build GRCh38
UniGene ID
Hs.377416
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024712.3 NP_078988.2 14 1486
GenBank mRNA AK023886.1 - 13 1250
AK056923.1 - 13 1449
AK300976.1 - 14 1500
BC015524.1 - 4 296
BC034410.1 - 11 1098

View Full Product Details