Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318007.1 | NP_001304936.1 | 3, 4 | 398 | |
NM_024792.2 | NP_079068.1 | 4, 5 | 600 | ||
GenBank mRNA | AF477201.1 | - | 4, 5 | 588 | |
AK025935.1 | - | 4, 5 | 539 | ||
AK125146.1 | - | 3, 4 | 805 | ||
AK292065.1 | - | 4, 5 | 539 | ||
BC009729.1 | - | 1, 2 | 93 | ||
BX389422.1 | - | 2, 3 | 145 |