Gene Transcripts

Gene Symbol
CCDC30
Entrez Gene ID
728621
Gene Name
coiled-coil domain containing 30
Gene Aliases
PFD6L, PFDN6L
Chromosome Location
Chr. 1: 42521240 - 42654664 on Build GRCh38
UniGene ID
Hs.729640
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK024625.1 - 2 164
AL832217.1 - 5 762
AY639646.1 - 1 245

View Full Product Details